missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEX5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | BEX5 |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18079335
|
Proteintech
25281-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18089475
|
Proteintech
25281-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BEX5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BEX5. This antibody recognizes human, mouse antigen. The BEX5 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| BEX5 | |
| Immunohistochemistry, ELISA | |
| Rabbit | |
| 340542 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 600 μg/mL | |
| Liquid | |
| Human, Mouse | |
| BEX5, brain expressed, X linked 5, NADE2, NGFRAP1 like protein 1, NGFRAP1L1, Protein BEX5 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title