Learn More
BEX5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25281-1-AP-150UL
 View more versions of this product
Product Code. 18079335
  • 4836.10 NOK / 150µL
Description
The BEX5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BEX5. This antibody recognizes human, mouse antigen. The BEX5 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
BEX5
600 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunohistochemistry, ELISA
BEX5, brain expressed, X linked 5, NADE2, NGFRAP1 like protein 1, NGFRAP1L1, Protein BEX5
150 μL
340542
Store at -20°C. Stable for one year after shipment.