missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech Beta Galactosidase Mouse anti-Human, Mouse, Rat, CoraLite 488, Clone: 4F4F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66586-100UL
Additional Details : Weight : 0.00010kg
Description
Beta Galactosidase Monoclonal antibody apecifically detetects Beta Galactosidase antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
Beta Galactosidase | |
Monoclonal | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
GLB1 | |
Beta Galactosidase fusion protein Ag8069 | |
100 μL | |
2720 | |
Store at -20°C. Avoid exposure to light. | |
IgG1 |
Immunofluorescence | |
4F4F4 | |
CoraLite 488 | |
BC007493 | |
Mouse | |
Protein G purification | |
Primary | |
Human, Mouse, Rat | |
Liquid |