missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Galactosidase Mouse anti-Human, Mouse, Rat, CoraLite 488, Clone: 4F4F4, Proteintech
Mouse Monoclonal Antibody
1651.35 NOK - 5307.92 NOK
Specifications
| Antigen | Beta Galactosidase |
|---|---|
| Clone | 4F4F4 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17701299
|
Proteintech
CL488-66586-20UL |
20 μL |
1651.35 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17659866
|
Proteintech
CL488-66586-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Beta Galactosidase Monoclonal antibody apecifically detetects Beta Galactosidase antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| Beta Galactosidase | |
| Immunofluorescence | |
| CoraLite 488 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 2720 | |
| Beta Galactosidase fusion protein Ag8069 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 4F4F4 | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| BC007493 | |
| GLB1 | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title