missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Galactosidase Mouse anti-Human, Mouse, Rat, CoraLite 488, Clone: 4F4F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66586-20UL
This item is not returnable.
View return policy
Description
Beta Galactosidase Monoclonal antibody apecifically detetects Beta Galactosidase antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| Beta Galactosidase | |
| Monoclonal | |
| 1000 ug/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| GLB1 | |
| Beta Galactosidase fusion protein Ag8069 | |
| 20 μL | |
| 2720 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Immunofluorescence | |
| 4F4F4 | |
| CoraLite 488 | |
| BC007493 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction