Learn More
Benzyl Phenyl Sulfide, Sigma-Aldrich™
Specifications
Specifications
| CAS | 831-91-4 |
| Melting Point | 40°C to 44°C |
| MDL Number | MFCD00003066 |
| Quantity | 25 g |
| Synonym | Benzyl phenyl sulfide, Benzyl phenyl sulfide |
| Molecular Weight (g/mol) | 200.3 |
| Grade | Research |