Learn More
BD Difco™ Adjuvants
Product Code. 11719062
Change view
Click to view available options
Product Type:
Complete H37Ra
Freund's Complete Adjuvant
Freund's Incomplete Adjuvant
Mycobacterium butyricum
Mycobacterium tuberculosis H37Ra
Quantity:
6 x 100 mg
6 x 10 mL
Unit Size:
600mg
60mL
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
11719062 Freund's Complete Adjuvant 6 x 10 mL 60mL
11729062 Freund's Incomplete Adjuvant 6 x 10 mL 60mL
17224363 Mycobacterium butyricum 6 x 100 mg Pack of 6
11762934 Complete H37Ra 6 x 10 mL Pack of 6
10218823 Mycobacterium tuberculosis H37Ra 6 x 100 mg 600mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Not intended for human or therapeutic use.
Product Code. 11719062 Supplier BD Supplier No. 263810
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Not intended for human or therapeutic use.
Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)
  • Adjuvants are mixed with aqueous antigen solution or suspension
  • Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
  • With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
  • Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in cool, dry conditions in well sealed containers, away from oxidizing agents.
For Use With (Application) Immunological studies with laboratory animals
Formulation Suspension of 5mg M. butyricum in a mixture of paraffin oil and an emulsifying agent
Quantity 6 x 10 mL
Product Type Freund's Complete Adjuvant