Learn More
BD Cytometric Bead Array (CBA) Mouse Enhanced Sensitivity Master Buffer Kit
Product Code. 15861749
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
Each
Product Code. Quantity unitSize
15861749 100 Tests Each
15881749 500 Tests Each
2 options
Product Code. 15861749
Brand: BD 562246
Includes
Assay Diluent (150mL), Capture Bead Diluent (30mL), Detection Reagent Diluent A (15mL), Detection Reagent Diluent B (35mL), Wash Buffer (650mL)
For use in Flow Cytometry
Host Species:
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Cross Reactivity Mouse
Includes Assay diluent 150mL, capture bead Diluent 30mL, detection reagent diluent A 15mL, detection reagent diluent B 35mL, Wash Buffer 650mL
Quantity 100 Tests
Content And Storage 2°C to 8°C
For Use With (Application) Flow Cytometry
Product Type Mouse enhanced sensitivity master Buffer kit