Learn More
BD Antibody Diluent
Product Code. 15879678
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
Each
Product Code. Quantity unitSize
15879678 125 mL Each
1 options
Product Code. 15879678
Brand: BD 559148
For acetone-fixed, frozen tissue sections and formalin-fixed, paraffin-embedded tissue sections
Premixed, ready to use, sodium phosphate-based buffer solution, useful for the dilution of primary and/or secondary antibodies to be used in immunohistochemistry applications
  • Storage buffer: Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide
Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibody diluent
Quantity 125 mL
For Use With (Application) Immunohistochemistry (IHC), Staining of Intra-cellular Antigens
Form IHCH
Content And Storage 4°C