Learn More
BCL6 Mouse anti-Human, Clone: 1E6B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66340-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody only recognize human species.
BCL6 is a zinc finger transcription factor and contains an N-terminal POZ domain. This protein acts as a sequence-specific repressor of transcription, and has been shown to modulate the transcription of START-dependent IL-4 responses of B cells. This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors.Specifications
| BCL6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P41182 | |
| BCL6 | |
| BCL6 Fusion Protein Ag15519 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1E6B1 | |
| Unconjugated | |
| BCL6 | |
| B cell CLL/lymphoma 6, B cell lymphoma 5 protein, B cell lymphoma 6 protein, BCL6, LAZ3, Protein LAZ 3, ZBTB27, Zinc finger protein 51, ZNF51 | |
| Mouse | |
| Protein G | |
| RUO | |
| 604 | |
| -20°C | |
| Liquid |