missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL6 Mouse anti-Human, Clone: 1E6B1, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | BCL6 |
|---|---|
| Clone | 1E6B1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858125
|
Proteintech
66340-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848125
|
Proteintech
66340-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody only recognize human species.
BCL6 is a zinc finger transcription factor and contains an N-terminal POZ domain. This protein acts as a sequence-specific repressor of transcription, and has been shown to modulate the transcription of START-dependent IL-4 responses of B cells. This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors.Specifications
| BCL6 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B cell CLL/lymphoma 6, B cell lymphoma 5 protein, B cell lymphoma 6 protein, BCL6, LAZ3, Protein LAZ 3, ZBTB27, Zinc finger protein 51, ZNF51 | |
| BCL6 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1E6B1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P41182 | |
| 604 | |
| BCL6 Fusion Protein Ag15519 | |
| Primary | |
| -20°C | |
| BCL6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title