missing translation for 'onlineSavingsMsg'
Learn More

BCL6 Mouse anti-Human, Clone: 1E6B1, Proteintech

Product Code. 16858125 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16858125 20 μL 20µL
16848125 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16858125

Brand: Proteintech 663401IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody only recognize human species.

BCL6 is a zinc finger transcription factor and contains an N-terminal POZ domain. This protein acts as a sequence-specific repressor of transcription, and has been shown to modulate the transcription of START-dependent IL-4 responses of B cells. This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors.

Specifications

Antigen BCL6
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1E6B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BCL6
Gene Accession No. P41182
Gene Alias B cell CLL/lymphoma 6, B cell lymphoma 5 protein, B cell lymphoma 6 protein, BCL6, LAZ3, Protein LAZ 3, ZBTB27, Zinc finger protein 51, ZNF51
Gene Symbols BCL6
Host Species Mouse
Immunogen BCL6 Fusion Protein Ag15519
Purification Method Protein G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 604
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.