missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl2 Mouse anti-Mouse, CoraLite 555, Clone: 1B5B9, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 5307.92 NOK
Specifications
| Antigen | Bcl2 |
|---|---|
| Clone | 1B5B9 |
| Applications | Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | CoraLite 555 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17711369
|
Proteintech
CL555-66799-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17699906
|
Proteintech
CL555-66799-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl2 Monoclonal antibody apecifically detetects Bcl2 antigen in Mouse Samples. It is validated for Immunofluorescence, Flow Cytometry.Specifications
| Bcl2 | |
| Immunofluorescence, Flow Cytometry | |
| CoraLite 555 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 12043 | |
| Bcl2 fusion protein Ag24261 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 1B5B9 | |
| Monoclonal | |
| Liquid | |
| Mouse | |
| NM-009741 | |
| Bcl2 | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title