missing translation for 'onlineSavingsMsg'
Learn More

Bcl2 Mouse anti-Mouse, CoraLite 555, Clone: 1B5B9, Proteintech

Product Code. 17711369 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17711369 20 μL 20µL
17699906 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17711369

Brand: Proteintech CL5556679920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bcl2 Monoclonal antibody apecifically detetects Bcl2 antigen in Mouse Samples. It is validated for Immunofluorescence, Flow Cytometry.

Specifications

Antigen Bcl2
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 1B5B9
Concentration 1000 ug/mL
Conjugate CoraLite 555
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. NM-009741
Gene Symbols Bcl2
Host Species Mouse
Immunogen Bcl2 fusion protein Ag24261
Purification Method Protein G purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 12043
Target Species Mouse
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.