missing translation for 'onlineSavingsMsg'
Learn More

BCAR1 Mouse anti-Human, Unconjugated, Clone: 2D11C10, Proteintech

Mouse Monoclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen BCAR1
Clone 2D11C10
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16869735
View Documents
Proteintech
67215-1-IG-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16859735
View Documents
Proteintech
67215-1-IG-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The BCAR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAR1. This antibody recognizes human antigen. The BCAR1 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.

Specifications

Specifications

BCAR1
1000 μg/mL
Monoclonal
Mouse
9564
Primary
2D11C10
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
Unconjugated
Human
IgG2a
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.