Learn More
BCAR1 Mouse anti-Human, Unconjugated, Clone: 2D11C10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67215-1-IG-150UL
 View more versions of this product
Product Code. 16859735
  • 4836.10 NOK / 150µL
Description
The BCAR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAR1. This antibody recognizes human antigen. The BCAR1 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
BCAR1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
2D11C10
Unconjugated
150 μL
9564
IgG2a