Learn More
Fisherbrand™ Alkaline Battery
Product Code. 15318754 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Battery Type:
9 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Battery Type Unit Size
15318754 9 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15318754
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
Use this 9V battery with a wide range of electronic devices.
  • 9V Alkaline Battery
Specifications
Battery Type 9 V
Quantity 1 Ea.
Voltage 9 V