Learn More
Brannan™ Batch Sampling Freezer Thermometer with Calibration Certificate
Product Code. 17863058
Click to view available options
Calibrated:
3 Point Traceable
3-point UKAS
Unit Size:
Each
Product Code. 17863058
Brand: Brannan™ 22/101/2 + 49/503/0
Only null left
Add to basket
Add to basket
Precision test batch sampling freezer thermometers sealed into a 30 mL polypropylene bottle containing a heat transfer medium, ideally suited to monitor the temperature of samples in a controlled storage. Complete with traceable calibration.
  • Non-hazardous LO-tox™ filling
  • Bottle should be filled with monpropylene glycol or other heat transfer medium before use
  • Temperature measurement unaffected by short term fluctuations
  • Supplied with calibration
  • Temperature range: -30°C to 0°C
  • Supplied with statement of accuracy
  • Made in UK
Specifications
Bottle Fill Customer to fill with monopropylene glycol or other heat transfer medium before use
Calibration Points -25°C, -15°C, -5°C
Certifications/Compliance ISO 17025 UKAS
For Use With (Application) Monitor temperature of samples in a controlled storage
Height (English) 5.71 in.
Height (Metric) 14.5 cm
Includes Polypropylene bottle and glass thermometer
Length (Metric) 14.5 cm
Material Glass
Thermometer Fill No
Weight (English) 1.9577 oz.
Weight (Metric) 55.5 g
Width (English) 0.14 in.
Max. Temperature (English) 32°F
Max. Temperature (Metric) 0°C
Min. Temperature (English) -22°F
Min. Temperature (Metric) -30°C
Temperature Range (English) -22°F to +32°F
Temperature Range (Metric) -30°C to 0°C
Calibrated 3-point UKAS
For Use With (Equipment) Freezer
Battery Type None
Accuracy ± 0.7°C
Subdivision 0.5°C
Display None
Width (Metric) 3.5 cm