Learn More
Elma Basket System
Product Code. 17848835
Click to view available options
Dimensions (L x W x H):
70 x 155 x 35 mm
105 x 110 x 50 mm
105 x 195 x 50 mm
105 x 195 x 95 mm
105 x 255 x 95 mm
105 x 455 x 55 mm
195 x 250 x 90 mm
255 x 450 x 55 mm
195 x 250 x 135 mm
255 x 280 x 135 mm
255 x 450 x 135 mm
Model:
Basket System Modular Type 10
Basket System Modular Type 100
Basket System Modular Type 120
Basket System Modular Type 150
Basket System Modular Type 180
Basket System Modular Type 20
Basket System Modular Type 30
Basket System Modular Type 300
Basket System Modular Type 40
Basket System Modular Type 60
Basket System Modular Type 80
Unit Size:
Each
Product Code. 17848835
Brand: Elma 1111838
Only null left
Add to basket
Add to basket
Modular basket system for Elmasonic P, Elmasonic Select, EASY
  • Stainless steel with plastic handles
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Type Modular Basket System
Capacity (Metric) 5.0 kg
Capacity (English) 11 lb.
For Use With (Equipment) Fisherbrand Select. Fisherbrand EASY. Elmasonic EASY. Elmasonic P
Dimensions (L x W x H) 105 x 455 x 55 mm
Model Basket System Modular Type 80