Learn More
Eppendorf™ White Glove Delivery per unit
Product Code. 15783685
Change view
Click to view available options
Service Offering:
White Glove Delivery (>180 kg)
White Glove Delivery (<100 kg)
White Glove Delivery (100 to 180 kg)
Unit Size:
Each
Product Code. Service Offering unitSize
15783685 White Glove Delivery (100 to 180 kg) Each
15773685 White Glove Delivery (<100 kg) Each
15793685 White Glove Delivery (>180 kg) Each
3 options
Product Code. 15783685
Brand: Eppendorf™ 2277000003
Delivery on the ground floor or on a different floor with freight elevator inkl. unpacking and taking back of packaging. Shipment with stair climb to be ordered when needed.
  • White glove delivery per unit
  • Delivery on the ground floor or on a different floor with freight elevator inkl. unpacking and taking back of packaging
  • Shipment with stair climb to be ordered when needed
TRUSTED_SUSTAINABILITY
Specifications
Service Offering White Glove Delivery (100 to 180 kg)