Learn More
B4GALT7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10535-1-AP-20UL
 View more versions of this product
Product Code. 16896973
  • 1710.33 NOK / 20µL
Description
Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.
Specifications
Specifications
B4GALT7
Polyclonal
Unconjugated
B4GALT7
B4GAL T7, B4GALT7, Beta 1,4 GalTase 7, beta4Gal T7, XGALT 1, XGALT1, XGPT, XGPT1
Rabbit
Antigen Affinity Chromatography
RUO
11285
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UBV7
B4GALT7
B4GALT7 Fusion Protein Ag0793