Learn More
B4GALT7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10535-1-AP-20UL
This item is not returnable.
View return policy
Description
Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.Specifications
| B4GALT7 | |
| Polyclonal | |
| Unconjugated | |
| B4GALT7 | |
| B4GAL T7, B4GALT7, Beta 1,4 GalTase 7, beta4Gal T7, XGALT 1, XGALT1, XGPT, XGPT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11285 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UBV7 | |
| B4GALT7 | |
| B4GALT7 Fusion Protein Ag0793 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |