missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALT7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | B4GALT7 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16896973
|
Proteintech
10535-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16886973
|
Proteintech
10535-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.Specifications
| B4GALT7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UBV7 | |
| 11285 | |
| B4GALT7 Fusion Protein Ag0793 | |
| Primary | |
| -20°C | |
| B4GALT7 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B4GAL T7, B4GALT7, Beta 1,4 GalTase 7, beta4Gal T7, XGALT 1, XGALT1, XGPT, XGPT1 | |
| B4GALT7 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title