Learn More
IBA Lifesciences Avidin
Product Code. 12893186 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12893186 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12893186 Supplier IBA Lifesciences Supplier No. 20204050
  • ONLINE EXCLUSIVE PRICE
    3935.00 NOK
    3585.79 NOK / 50mg
    Save 349.21 NOK
    8% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Avidin can efficiently block of biotin and biotinlyated proteins.
Avidin is a tetrameric biotin binding protein (KD ∼1 x 10-14) extracted from egg white. It prevents biotin and biotinylated proteins from binding to Strep-Tactin™ or Strep-Tactin™XT, whereas binding of the Twin-Strep-tag™ and Strep-tag™II is not influenced.
Biotin and/or biotinylated proteins can potentially reduce the binding capacity of Strep-Tactin™ as well as Strep-Tactin™XT and are present in cell lysates and cell culture media, especially from mammalian or insect cells. Thus, if Twin-Strep-tag™ and Strep-tag™II fusion proteins from biotin-containing extracts or media are intended to be purified via Strep-Tactin™ or Strep-Tactin™XT affinity chromatography, biotin has to be masked by addition of avidin prior to application onto the resin.
Specifications
Quantity 50 mg
For Research Use Only.