Learn More
MP Biomedicals™ Atherogenic Diet
Shop All MP Biomedicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
10 kg
20 kg
50 kg
missing translation for 'unitSize'
10kg
20kg
50kg
Beschreibung
- Atherogenic diet is rich in cholesterol and other atherogenic factors
- Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease
Spezifikation
Spezifikation
| Product Type | Animal Diet |
| For Use With (Application) | Rodent diet |
| Quantity | 10 kg |
| Storage Requirements | 2°C to 8°C |
Atherogenic diet is rich in cholesterol and other atherogenic factors. Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur