Learn More
ATG5 (Autophagy Marker) Mouse, Clone: ATG5/2101, enQuireBio™
Mouse Monoclonal Antibody
2058.63 NOK - 5275.00 NOK
Specifications
Antigen | ATG5 (Autophagy Marker) |
---|---|
Clone | ATG5/2101 |
Concentration | 200μg/mL |
Applications | ELISA, Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951238
|
EnquireBio
9474-MSM1-P1 |
100 μg |
5275.00 NOK
100µg |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941238
|
EnquireBio
9474-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
ATG5 (Autophagy Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human ATG5 protein (apprx. aa 1-119) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
50-32kDa |
ATG5/2101 | |
ELISA, Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
9474 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cytoplasmic | |
THP-1, PANC-1, Raji or HeLa cells. Brain, Ovary, Endometrium, Colon or Duodenum |