missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASF1B Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813126
Additional Details : Weight : 0.05000kg
Description
ASF1B Monoclonal antibody specifically detects ASF1B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
ASF1B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9DAP7, Q9NVP2 | |
Mouse | |
Protein A/G | |
RUO | |
304648, 55723, 66929 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI1H5 | |
Unconjugated | |
ASF1B | |
1700003K02Rik; AA409591; anti-silencing function 1B histone chaperone; anti-silencing function protein 1 homolog B; ASF1 anti-silencing function 1 homolog B; Asf1b; CCG1-interacting factor A-II; CIA-II; hAsf1; hAsf1b; hCIA-II; histone chaperone ASF1B; mCIA-II | |
Full length human recombinant protein of human ASF1B (NP_060624) produced in E.coli. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |