missing translation for 'onlineSavingsMsg'
Learn More

ASF1B Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene

Product Code. p-7225517
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17557834 30 μL 30µL
17537834 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17557834

Brand: Origene Technologies TA813126S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASF1B Monoclonal antibody specifically detects ASF1B in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ASF1B
Applications Western Blot
Classification Monoclonal
Clone OTI1H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ASF1B
Gene Accession No. Q9DAP7, Q9NVP2
Gene Alias 1700003K02Rik; AA409591; anti-silencing function 1B histone chaperone; anti-silencing function protein 1 homolog B; ASF1 anti-silencing function 1 homolog B; Asf1b; CCG1-interacting factor A-II; CIA-II; hAsf1; hAsf1b; hCIA-II; histone chaperone ASF1B; mCIA-II
Host Species Mouse
Immunogen Full length human recombinant protein of human ASF1B (NP_060624) produced in E.coli.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304648, 55723, 66929
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.