missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Mouse anti-Human, Mouse, Rat, Rabbit, Clone: 1C10E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68069-1-IG-150UL
This item is not returnable.
View return policy
Description
ARF1 Monoclonal antibody apecifically detetects ARF1 antigen in Human, Mouse, Rat, Rabbit Samples. It is validated for Western Blot, Immunofluorescence, ELISA.Specifications
| ARF1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| ARF1 | |
| ARF1 fusion protein Ag28021 | |
| 150 μL | |
| 375 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG1 |
| Western Blot, Immunofluorescence, ELISA | |
| 1C10E3 | |
| Unconjugated | |
| BC009247 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction