missing translation for 'onlineSavingsMsg'
Learn More

ARF1 Mouse anti-Human, Mouse, Rat, Rabbit, Clone: 1C10E3, Proteintech

Product Code. p-7225929 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17629746 20 μL 20µL
17619746 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17629746 Supplier Proteintech Supplier No. 680691IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARF1 Monoclonal antibody apecifically detetects ARF1 antigen in Human, Mouse, Rat, Rabbit Samples. It is validated for Western Blot, Immunofluorescence, ELISA.

Specifications

Antigen ARF1
Applications Western Blot, Immunofluorescence, ELISA
Classification Monoclonal
Clone 1C10E3
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC009247
Gene Symbols ARF1
Host Species Mouse
Immunogen ARF1 fusion protein Ag28021
Purification Method Protein G purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 375
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.