missing translation for 'onlineSavingsMsg'
Learn More

ARF1 Mouse anti-Human, Mouse, Rat, Rabbit, Clone: 1C10E3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68069-1-IG-20UL

 View more versions of this product

Product Code. 17629746

  • 1710.33 NOK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ARF1 Monoclonal antibody apecifically detetects ARF1 antigen in Human, Mouse, Rat, Rabbit Samples. It is validated for Western Blot, Immunofluorescence, ELISA.
Specifications

Specifications

ARF1
Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
ARF1
ARF1 fusion protein Ag28021
20 μL
375
Store at -20°C. Stable for one year after shipment.
IgG1
Western Blot, Immunofluorescence, ELISA
1C10E3
Unconjugated
BC009247
Mouse
Protein G purification
Primary
Human, Mouse, Rat, Rabbit
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.