Learn More
Miele Professional™ Appliance lid
Shop All Miele Products
Click to view available options
:
Lid; Dimensions (D x W):760 x 90mm
Description
To cover the top of a machine
Specifications
Specifications
| Material | Stainless Steel |
| Product Type | Cover |
| Color | Stainless Steel |
| For Use With (Equipment) | Washer-disinfectors, laboratory |