missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ APOC1 (Human) Recombinant Protein

Human APOC1 full-length ORF ( AAH09698, 1 a.a. - 83 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000341-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16151711

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MRLFLSLPVLVVVLSIVLEGPAPAQGTPDVSSALDKLKEFGNTLEDKARELISRIKQSELSAKMREWFSETFQKVKEKLKIDS
Specifications

Specifications

AAH09698
apolipoprotein C-I
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
341
Wheat germ expression system
25 μg
APOC1
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers