missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ APOC1 (Human) Recombinant Protein
Human APOC1 full-length ORF ( AAH09698, 1 a.a. - 83 a.a.) recombinant protein with GST-tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH09698 |
---|---|
Gene ID (Entrez) | 341 |
Name | apolipoprotein C-I |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141711
|
Abnova™
H00000341-P01.10UG |
10 μg |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16151711
|
Abnova™
H00000341-P01.25UG |
25 μg |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MRLFLSLPVLVVVLSIVLEGPAPAQGTPDVSSALDKLKEFGNTLEDKARELISRIKQSELSAKMREWFSETFQKVKEKLKIDS
Specifications
AAH09698 | |
apolipoprotein C-I | |
125% SDS-PAGE Stained with Coomassie Blue | |
APOC1 | |
GST |
341 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |