missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ APOC1 (Human) Recombinant Protein
Human APOC1 full-length ORF ( AAH09698, 1 a.a. - 83 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000341-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MRLFLSLPVLVVVLSIVLEGPAPAQGTPDVSSALDKLKEFGNTLEDKARELISRIKQSELSAKMREWFSETFQKVKEKLKIDS
Specifications
AAH09698 | |
apolipoprotein C-I | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
341 | |
Wheat germ expression system | |
10 μg | |
APOC1 | |
GST |