missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antizyme inhibitor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
| Antigen | Antizyme inhibitor 1 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Antizyme inhibitor (AZIN1) protein that positively regulates ornithine decarboxylase (ODC) activity and polyamine uptake. AZI is an enzymatically inactive ODC homolog that counteracts the negative effect of ODC antizymes (AZs) OAZ1, OAZ2 and OAZ3 on ODC activity by competing with ODC for antizyme-binding. Inhibits antizyme-dependent ODC degradation and releases ODC monomers from their inactive complex with antizymes, leading to formation of the catalytically active ODC homodimer and restoring polyamine production.Spécification
| Antizyme inhibitor 1 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| O14977, O35484, Q63764 | |
| 51582, 54375, 58961 | |
| Antizyme inhibitor 1 Fusion Protein Ag2082 | |
| Primary | |
| -20°C | |
| AZIN1 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Antizyme inhibitor 1, AZI, AZIN1, OAZI, OAZIN, ODC1L | |
| AZIN1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit