missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antizyme inhibitor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11548-1-AP-20UL
This item is not returnable.
View return policy
Description
Antizyme inhibitor (AZIN1) protein that positively regulates ornithine decarboxylase (ODC) activity and polyamine uptake. AZI is an enzymatically inactive ODC homolog that counteracts the negative effect of ODC antizymes (AZs) OAZ1, OAZ2 and OAZ3 on ODC activity by competing with ODC for antizyme-binding. Inhibits antizyme-dependent ODC degradation and releases ODC monomers from their inactive complex with antizymes, leading to formation of the catalytically active ODC homodimer and restoring polyamine production.Specifications
| Antizyme inhibitor 1 | |
| Polyclonal | |
| Unconjugated | |
| AZIN1 | |
| Antizyme inhibitor 1, AZI, AZIN1, OAZI, OAZIN, ODC1L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51582, 54375, 58961 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14977, O35484, Q63764 | |
| AZIN1 | |
| Antizyme inhibitor 1 Fusion Protein Ag2082 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction