Learn More
Melford Antipain
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Reversible cysteine and serine protease inhibitor of cathepsins A and B, papain and trypsin.
Reversible cysteine and serine protease inhibitor of cathepsins A and B, papain and trypsin.
Working concentration: 1-100uM.
Specifications
Specifications
| CAS | 9087-70-1 |
| Molecular Formula | C27H44N10O6-2 HCl |
| Quantity | 5 mg |