Learn More
Molecular Probes™ Antibody Conjugate Purification Kit, 20-50 μg conjugate
Product Code. 15401064
Change view
Click to view available options
Quantity:
3 Purifications
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15401064 3 Purifications 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15401064 Supplier Molecular Probes™ Supplier No. A33087
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Antibody Conjugate Purification Kit, 20-50 μg conjugate
Molecular Probes™ Antibody Conjugate Purification kits efficiently purify antibody conjugates from unconjugated dye or biotin. This particular kit is optimized for the purification of 20–50 μg of antibody conjugate. Purification is accomplished using an easy-to-assemble size-exclusion spin column containing BioGel™ P-6 fine resin. Each kit contains sufficient material for 3 separate purifications, and can also be used to purify proteins with a molecular weight greater than 12 kDa away from unconjugated labels.
Additional Features of this Antibody Conjugate Purification Kit:
  • Purified conjugate ready-to-use in ∼10 minutes with ∼5 minutes of hands-on time
  • Designed for low-molecular weight labels (less than 3000 Da, e.g. Alexa Fluor™ dyes); cannot be used for removing unconjugated high-molecular weight labels (>8 kDa) such as enzymes (HRP) or proteins (R-phycoerythrin)
More about Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental set-up. To learn more about our labeling kits, see Kits for Labeling Proteins and Nucleic Acids in The Molecular Probes™ Handbook or visit our antibody labeling page.
Related Links:
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibody Conjugate Purification Kit
Quantity 3 Purifications
Content And Storage Store in refrigerator (2-8 °C).
Format Spin Column