Learn More
BD HiCK-3 Human Cytokine Positive Control Cells
Product Code. 15847988 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15847988 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15847988 Supplier BD Supplier No. 555063
 Restricted Item
Estimated Shipment: 02-12-2026
  • This suspension contains Human intracellular CytoKine-3 (HiCK-3) Positive Control Cells
  • The HiCK-3 frozen cell suspension contains fixed, non-permeabilized human lymphoid cells
  • The suspension includes cells that express detectable levels of intracellular IL-1α, IL-1β, IL-6, IL-12 p40/p70, IL-12 p70, and TNF as determined by immunofluorescent intracellular cytokine staining and flow cytometry
  • HiCK-3 cells contain a measurable proportion of cytokines
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5 x106 cells/mL
For Use With (Application) Intracellular staining (flow cytometry)
Quantity 1 mL