Learn More
Human IL-12/IL-23 p40 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 7 publications
1930.00 NOK - 20815.00 NOK
Specifications
| Antigen | IL-12/IL-23 p40 |
|---|---|
| Clone | 24901 |
| Concentration | LYOPH |
| Dilution | Neutralization 0.02-0.1 ug/mL |
| Applications | Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782538
![]() |
R&D Systems
MAB1510-SP |
25 μg |
1930.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744279
![]() |
R&D Systems
MAB1510-100 |
100 μg |
5145.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763838
![]() |
R&D Systems
MAB1510-500 |
500 μg |
20815.00 NOK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Human samples. It is validated for Neutralization.Specifications
| IL-12/IL-23 p40 | |
| LYOPH | |
| Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3593 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 p40/p35 heterodimer | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-12 (p40/p35 heterodimer), IL-23 (p40/p19 heterodimer), and p40 homodimers in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) IL-12 p40 or rpIL-12. |
| 24901 | |
| Neutralization 0.02-0.1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 | |
| IL12B | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only