missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A2 Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-11256-20UL
Additional Details : Weight : 0.00010kg
Description
Annexin A2 Polyclonal antibody apecifically detetects Annexin A2 antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.Specifications
Annexin A2 | |
Polyclonal | |
CoraLite 594 | |
BC016774 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Flow Cytometry | |
1000 ug/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
ANXA2 | |
Annexin A2 fusion protein Ag1777 | |
20 μL | |
302 | |
Store at -20°C. Avoid exposure to light. | |
IgG |