missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A2 Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1565.79 NOK - 4225.34 NOK
Specifications
Antigen | Annexin A2 |
---|---|
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | CoraLite 594 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17741379
|
Proteintech
CL594-11256-20UL |
20 μL |
1565.79 NOK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17649926
|
Proteintech
CL594-11256-100UL |
100 μL |
4225.34 NOK
100µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Annexin A2 Polyclonal antibody apecifically detetects Annexin A2 antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.Specifications
Annexin A2 | |
Polyclonal | |
Liquid | |
Human, Mouse, Rat | |
BC016774 | |
ANXA2 | |
IgG | |
Antigen affinity purification |
Flow Cytometry | |
CoraLite 594 | |
Rabbit | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
302 | |
Annexin A2 fusion protein Ag1777 | |
Primary | |
Store at -20°C. Avoid exposure to light. |