missing translation for 'onlineSavingsMsg'
Learn More

Annexin A2 Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech CL594-11256-100UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17649926

  • 4225.34 NOK / 100µL
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Annexin A2 Polyclonal antibody apecifically detetects Annexin A2 antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.
Specifications

Specifications

Annexin A2
Polyclonal
CoraLite 594
BC016774
Rabbit
Antigen affinity purification
Primary
Human, Mouse, Rat
Liquid
Flow Cytometry
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
ANXA2
Annexin A2 fusion protein Ag1777
100 μL
302
Store at -20°C. Avoid exposure to light.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers