Learn More
Ammonium aquopentachlororhodate(III), Rh 30% min
Product Code. 11347487
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11347487
Brand: Thermo Scientific Alfa Aesar 012645.03
Request Bulk or Custom Format
CAS: 63771-33-5 | Cl5H10N2ORh | 334.25 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63771-33-5
Molecular Formula Cl5H10N2ORh
Molecular Weight (g/mol) 334.25
MDL Number MFCD00064673
InChI Key SSZNXYVCMKGDBP-UHFFFAOYSA-K
Synonym diammonium aquapentachlororhodate, diammonium pentachlororhodiumdiuide hydrate
Specifications
Melting Point 210°C to 230°C (decomposition)
Physical Form Powder
Quantity 1 g
Formula Weight 334.26
Concentration or Composition (by Analyte or Components) Rh 30% min.
Chemical Name or Material Ammonium aquopentachlororhodate(III)