CAS RN 63771-33-5
CAS RN 63771-33-5
IUPAC Name:
diammonium pentachlororhodiumdiuide hydrate
Synonyms:
diammonium pentachlororhodiumdiuide hydrate
Molecular Weight (g/mol):
334.25
Molecular Formula:
Cl5H10N2ORh
InChi Key:
SSZNXYVCMKGDBP-UHFFFAOYSA-K
SMILES:
[NH4+].[NH4+].O.Cl[Rh--](Cl)(Cl)(Cl)Cl
1
–
1
of
1
results
1
Ammonium aquopentachlororhodate(III), Rh 30% min
CAS: 63771-33-5 Molecular Formula: Cl5H10N2ORh Molecular Weight (g/mol): 334.25 MDL Number: MFCD00064673 InChI Key: SSZNXYVCMKGDBP-UHFFFAOYSA-K Synonym: diammonium aquapentachlororhodate,diammonium pentachlororhodiumdiuide hydrate