missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMH Monoclonal Antibody (C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA543971
Additional Details : Weight : 0.01000kg
Description
Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.Specifications
AMH | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P03971, P79295 | |
AMH | |
Thr118-Arg134, with N-terminal His Tag | |
100 μL | |
Primary | |
Human, Porcine, Zebrafish | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot | |
C6 | |
Unconjugated | |
AMH | |
AMH; Anti - Mullerian hormone (Mulerian inhibiting substance); anti-Muellerian hormone; anti-Mullerian hormone; MIF; MIS; Muellerian-inhibiting factor; muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance | |
Mouse | |
Protein A/G | |
RUO | |
268, 397578, 493624 | |
-20°C | |
Liquid |