missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AMH Monoclonal Antibody (C6), Invitrogen™

Product Code. p-7240805
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17664426 20 μL 20µL
17654426 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17664426

Brand: Invitrogen™ MA543972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene AMH
Gene Accession No. P03971
Gene Alias AMH; Anti - Mullerian hormone (Mulerian inhibiting substance); anti-Muellerian hormone; anti-Mullerian hormone; MIF; MIS; Muellerian-inhibiting factor; muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance
Gene Symbols AMH
Host Species Mouse
Immunogen Thr118-Arg134, with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268, 493624
Target Species Human, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.