Learn More
Cytiva Amersham™ ECL Advance™ Blocking Reagent
Product Code. 10106424 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
40g
Unit Size:
40g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10106424 40g 40g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10106424 Supplier Cytiva Supplier No. RPN418
  • ONLINE EXCLUSIVE PRICE
    1170.00 NOK
    1099.01 NOK / 40g
    Save 70.99 NOK
    6% Off
 Restricted Item
In Stock
Use for Controlling background noise. Cytiva Life Sciences™ Amersham™ ECL Advance™ is Blocking Reagent controls background noise while retaining a high level of signal intensity.
Fully compatible with both Hybond™ ECL™ (nitrocellulose) and Hybond-P (PVDF) membranes.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For blocking Nitrocellulose and PVDF membranes prior to detection with Amersham ECL, Amersham ECL Prime or Amersham ECL Select western blotting detection reagents
Content And Storage 2° to 8°C
Product Type Blocking reagent
Quantity 40g
Product Line Amersham