Learn More
Cytiva Amersham™ CyDye™ Antibody Labeling Kits
Product Code. 10754167 Shop All Cytiva Products
Click to view available options
Label or Dye:
Cy2 Reactive Dye
Cy2 mono-Functional Dye
Cy3 Reactive Dye
Cy5 Reactive Dye
Product Type:
Cy2 Ab Labeling Kit
Cy2 MAb Labeling Kit
Cy3 Ab Labeling Kit
Cy5 Ab Labeling Kit
Unit Size:
1 set
Product Code. 10754167
Brand: Cytiva PA33000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Convenient way to tag antibodies with CyDye fluorescent dyes
CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
  • Conjugations are carried out under mild conditions and are complete within 1 hour
  • Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings
Order Info
Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For the conjugation of proteins with Cy3 Reactive Dye
Quantity Each
Product Type Cy3 Ab Labeling Kit
pH Range 7 to 7.3
Molecular Weight (g/mol) 949.11
Solubility Cy3 dye: Soluble in cold water and hot water Coupling buffer: Soluble in cold water and hot water Elution buffer: Soluble in cold water and hot water
Content And Storage 2°C to 8°C, protected from light
Label or Dye Cy3 Reactive Dye
Color Red
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1