Learn More
AM966, MedChemExpress
Product Code. 16422527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16422527 50 mg 50mg
16476706 100 mg 100mg
16456418 10 mg 10mg
16439007 5 mg 5mg
4 options
Product Code. 16422527
Brand: Medchem Express HY15277
MedChemExpress AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).
AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).
Specifications
Chemical Name or Material AM966
CAS 1228690-19-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H23ClN2O5
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (203.70 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4Cl)C)=O)C(C)=NO3)C=C2)C=C1
Molecular Weight (g/mol) 490.93
Formula Weight 490.93
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation